Mercury standard solution, 10 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Specpure®
45259.AEEA 101 EUR
45259.AE
Mercury standard solution, 10 mg/L Hg in 5% nitric acid (from Hg), standard for ICP, Specpure®
Standards Cationic Standards Mercury Standards
Molecular Formula: Hg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035

Certifikace: Traceable to NIST
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR