Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid (from ZrOCl₂), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Specpure®
88119.AEEA 42 EUR
88119.AE 88119.AP
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid (from ZrOCl₂), standard for AAS, Specpure®
Standards Cationic Standards Zirconium Standards
Molecular Formula: ZrCl₂O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011303
Certifikace: Traceable to NIST