2-Ethylhexyl salicylate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Octyl Salicylate, Salicylic acid 2-Ethylhexyl ester, 2-Ethylhexyl 2-hydroxybenzoate, Salicylic acid Octyl ester

M-Clarity™ quality level = MQ300

W514500-10KG-K W514500-1KG-K
SIALW514500-10KG-KEA 589 EUR
SIALW514500-10KG-K SIALW514500-1KG-K
2-Ethylhexyl salicylate, Sigma-Aldrich®
2-Ethylhexyl salicylate
Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
MDL Number: MFCD00053300
CAS Number: 118-60-5
EINECS: 204-263-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR