8-Bromo-2-tetralone

Supplier: Thermo Scientific

Synonyms: 8-Bromo-3,4-dihydronaphthalen-2(1H)-one

RH01477.250 RH01477.1
MAYBRH01477.250EA 0 EUR
MAYBRH01477.250 MAYBRH01477.1
8-Bromo-2-tetralone
8-Bromo-2-tetralone
Formula: C₁₀H₉BrO
MDL Number: MFCD03093390
CAS Number: 117294-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR