5-Methoxy-1-tetralone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Methoxy-3,4-dihydronaphthalenone

M-Clarity™ quality level = MQ100

113115-25G 113115-5G
SIAL113115-25GEA 302 EUR
SIAL113115-25G SIAL113115-5G
5-Methoxy-1-tetralone, Sigma-Aldrich®
5-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD00001692
CAS Number: 33892-75-0
EINECS: 251-723-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR