7-Methoxy-1-tetralone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 7-Methoxy-3,4-dihydronaphthalen-1(2H)-one

M-Clarity™ quality level = MQ100

163368-25G 163368-5G
SIAL163368-25GEA 624 EUR
SIAL163368-25G SIAL163368-5G
7-Methoxy-1-tetralone, Sigma-Aldrich®
7-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD00001696
CAS Number: 6836-19-7
EINECS: 229-916-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR