5,8-Dimethoxy-1-tetralone, Sigma-Aldrich®

Supplier: Merck

569658-1G 569658-5G
SIAL569658-1GEA 399 EUR
SIAL569658-1G SIAL569658-5G
5,8-Dimethoxy-1-tetralone, Sigma-Aldrich®
5,8-Dimethoxy-1-tetralone
Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
CAS Number: 1015-55-0
EINECS: 213-803-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR