5,6,7,8-Tetrahydro-2-naphthol

Supplier: Thermo Scientific

BTB10211.1 BTB10211.10 BTB10211.50
MAYBBTB10211.1EA 0 EUR
MAYBBTB10211.1 MAYBBTB10211.10 MAYBBTB10211.50
5,6,7,8-Tetrahydro-2-naphthol
5,6,7,8-Tetrahydro-2-naphthol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001738
CAS Number: 1125-78-6
EINECS: 214-410-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR