trans,trans-2,4-Decadienal, Sigma-Aldrich®
Supplier: Merck
Synonyms: Decadienal, (2E,4E)-Deca-2,4-dienal, DDA
M-Clarity™ quality level = MQ300
W313505-100G-K W313505-1KG-K W313505-5KG-K
SIALW313505-100G-KEA 303 EUR
SIALW313505-100G-K SIALW313505-1KG-K SIALW313505-5KG-K
trans,trans-2,4-Decadienal, Sigma-Aldrich®
trans,trans-2,4-Decadienal
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00007007
CAS Number: 25152-84-5
EINECS: 246-668-9