3-Chloro-6-hydrazinopyridazine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Chloro-6-hydrazinylpyridazine

M-Clarity™ quality level = MQ100

632619-1G 632619-5G
SIAL632619-1GEA 27.1 EUR
SIAL632619-1G SIAL632619-5G
3-Chloro-6-hydrazinopyridazine, Sigma-Aldrich®
3-Chloro-6-hydrazinopyridazine
Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD00051740
CAS Number: 17284-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR