2-Chloro-4-methoxypyrimidine, Sigma-Aldrich®

Supplier: Merck

536458-1G 536458-5G
SIAL536458-1GEA 26.1 EUR
SIAL536458-1G SIAL536458-5G
2-Chloro-4-methoxypyrimidine, Sigma-Aldrich®
2-Chloro-4-methoxypyrimidine
Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD00194055
CAS Number: 22536-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR