2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%
Supplier: Thermo Scientific
Warning
Synonyms: N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide
135440500 135440100
ACRO135440500EA 257 EUR
ACRO135440500 ACRO135440100
2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%
2-Chloro-N-(2,6-dimethylphenyl)acetamide
Formula: C₁₀H₁₂ClNO
MW: 197.66 g/mol
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00000926
CAS Number: 1131-01-7
EINECS: 214-460-7
Order Now
Specification Test Results
Appearance White to brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥97.5 %