Potassium methyl malonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

63405-250G 63405-50G
SIAL63405-250GEA 172 EUR
SIAL63405-250G SIAL63405-50G
Potassium methyl malonate, Sigma-Aldrich®
Potassium methyl malonate
Formula: C₄H₅KO₄
MW: 156.18 g/mol
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR