Nitrilotriacetic acid disodium salt, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
N0128-100G
N0128-50G
SIALN0128-100GEA
145
EUR
SIALN0128-100G
SIALN0128-50G
Nitrilotriacetic acid disodium salt, Sigma-Aldrich®
Nitrilotriacetic acid disodium salt
Formula:
C₆H₇NNa₂O₆ |
MDL Number:
MFCD00064230 CAS Number: 15467-20-6 EINECS: 239-484-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...