Nitrilotriacetic acid trisodium salt, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

N0253-1KG N0253-250G
SIALN0253-1KGEA 137 EUR
SIALN0253-1KG SIALN0253-250G
Nitrilotriacetic acid trisodium salt, Sigma-Aldrich®
Nitrilotriacetic acid trisodium salt
Formula: C₆H₆NNa₃O₆
MW: 257.09 g/mol
MDL Number: MFCD00064231
CAS Number: 5064-31-3
EINECS: 225-768-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR