trans-Aconitic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid
M-Clarity™ quality level = MQ200
122750-100G 122750-25G
SIAL122750-100GEA 169 EUR
SIAL122750-100G SIAL122750-25G
trans-Aconitic acid, Sigma-Aldrich®
trans-Aconitic acid
Formula: C₆H₆O₆
MW: 174.11 g/mol
MDL Number: MFCD00002721
CAS Number: 4023-65-8
EINECS: 223-688-6