Ethylaluminium dichloride 1 mol/l in hexane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
251615-100ML 251615-800ML
SIAL251615-100MLEA 86.7 EUR
SIAL251615-100ML SIAL251615-800ML
Ethylaluminium dichloride 1 mol/l in hexane, Sigma-Aldrich®
Ethylaluminium dichloride
Formula: C₂H₅AlCl₂
MW: 126.95 g/mol
MDL Number: MFCD00000457
CAS Number: 563-43-9
EINECS: 209-248-6
UN: 3394
ADR: 4.2,I