(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 99%, ChiPros®
Supplier: Thermo Scientific
Warning
Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-amine
L17022.06 L17022.14 L17022.03
L17022.06EA 70.2 EUR
L17022.06 L17022.14 L17022.03
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine ≥99%, ee 99%, ChiPros®
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
Purity: 99+% CAS No.: 23357-52-0 Molecular Formula: C10H13N Formula Weight: 147.22 Flash Point: 110°(230°F) Refractive Index: 1.5645 Boiling Point: 118-120°/8mm Beilstein Reference: 2360279 MDL No.: MFCD00671630
Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 118…120 °C (8 mmHg)
Density: 1.035
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671630
CAS Number: 23357-52-0