2-Phenyl-1,3-dithiane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

279617-25G 279617-5G
SIAL279617-25GEA 262 EUR
SIAL279617-25G SIAL279617-5G
2-Phenyl-1,3-dithiane, Sigma-Aldrich®
2-Phenyl-1,3-dithiane
Formula: C₁₀H₁₂S₂
MW: 196.34 g/mol
MDL Number: MFCD00006656
CAS Number: 5425-44-5
EINECS: 226-568-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR