uvex pheos Safety Spectacles
Supplier: UVEX
9192.215 9192.225 9192.725 9192.785 9192.881 9192.385 9192.485 9192.745 9192.285 9192.720 9192.080
111-1245EA 29.6 EUR
111-1245 111-1246 111-1248 111-1252 111-1259 111-1247 111-1692 111-1877 111-1876 UVEX9192.720 UVEX9192.080
uvex pheos Safety Spectacles
Safety Glasses
The supravision HC-AF coating technology makes the uvex pheos lens highly scratch resistant on the outside and permanently anti-fog on the inside.
  • Provides 100% UV protection
  • Integral brow guard for additional coverage
  • Enhanced ventilation
  • High performance hinges
  • PPE compatibility (earmuffs, safety helmets and bump caps)
Certifikace: EN 166, EN 170