L(+)-Arabinose, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
A3256-100G A3256-10MG A3256-1KG A3256-25G A3256-500G
SIALA3256-100GEA 204 EUR
SIALA3256-100G SIALA3256-10MG SIALA3256-1KG SIALA3256-25G SIALA3256-500G
L(+)-Arabinose, Sigma-Aldrich®
L(+)-Arabinose
Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 158…160 °C
Density: 1.585 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
EINECS: 226-214-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM