3,5-Bis(trifluoromethyl)benzyl alcohol
Supplier: Thermo Scientific
Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanol
TL00661.1 TL00661EA TL00661.50
MAYBTL00661.1EA 0 EUR
MAYBTL00661.1 MAYBTL00661EA MAYBTL00661.50
3,5-Bis(trifluoromethyl)benzyl alcohol
3,5-Bis(trifluoromethyl)benzyl alcohol
Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 100 °C (14 mmHg)
Melting Pt: 53…54 °C
MDL Number: MFCD00009907
CAS Number: 32707-89-4
EINECS: 251-168-9