Isoniazid
Supplier: Thermo Scientific
|
Warning
|
BTB00366.1
BTB00366.10
MAYBBTB00366.1EA
0
EUR
MAYBBTB00366.1
MAYBBTB00366.10
Isoniazid
Isoniazid
Formula:
C₆H₇N₃O Melting Pt: 169…172 °C Density: 1.42 g/cm³ (20 °C) Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006426 CAS Number: 54-85-3 EINECS: 200-214-6 Merck Index: 13,05205 |