Hydrogen chloride 4 M in 1,4-dioxane 99%
Supplier: Thermo Scientific
Danger
A16935.JV A16935.JT A16935.JQ
A16935.JVEA 370 EUR
A16935.JV A16935.JT A16935.JQ
Hydrogen chloride 4 M in 1,4-dioxane 99%
Hydrogen chloride
Purity: 99% CAS No.: 7647-01-0 Molecular Formula: ClH Formula Weight: 36.46 Flash Point: 17°(63°F) MDL No.: MFCD00011324
Formula: HCl
MW: 36.46 g/mol
Density: 1.05
Flash Pt: 17 °C (63 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3,II