N-(3,4-Dichlorophenyl)-2-chloroacetamide
Supplier: Thermo Scientific
Synonyms: 2-Chloro-N-(3,4-dichlorophenyl)acetamide
SPB07409.1 SPB07409.10 SPB07409.50
MAYBSPB07409.1EA 0 EUR
MAYBSPB07409.1 MAYBSPB07409.10 MAYBSPB07409.50
N-(3,4-Dichlorophenyl)-2-chloroacetamide
N-(3,4-Dichlorophenyl)-2-chloroacetamide
Formula: C₈H₆Cl₃NO
MW: 238.498 g/mol
MDL Number: MFCD00032563
CAS Number: 20149-84-2