Bis(2-chloroethyl)ammonium chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Bis(2-chloroethyl)amine hydrochloride

M-Clarity™ quality level = MQ200

B38503-100G B38503-250G B38503-25G
SIALB38503-100GEA 60.3 EUR
SIALB38503-100G SIALB38503-250G SIALB38503-25G
Bis(2-chloroethyl)ammonium chloride, Sigma-Aldrich®
Bis(2-chloroethyl)ammonium chloride
Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
MDL Number: MFCD00012515
CAS Number: 821-48-7
EINECS: 212-479-5
UN: 2923
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR