2-Chloro-4-fluorophenylacetonitrile ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 2-(2-Chloro-4-fluorophenyl)acetonitrile
B23132.06 B23132.14
B23132.06EA 32.9 EUR
B23132.06 B23132.14
2-Chloro-4-fluorophenylacetonitrile ≥97%
2-Chloro-4-fluorophenylacetonitrile
Purity: 97% CAS No.: 75279-56-0 Molecular Formula: C8H5ClFN Formula Weight: 169.59 Melting Point: 67-69° UN No.: 3439 MDL No.: MFCD00236029
Formula: C₈H₅ClFN
MW: 169.59 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00236029
CAS Number: 75279-56-0
UN: 3439
ADR: 6.1,II