2-Chloro-3-methoxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
655244-1G
655244-5G
SIAL655244-1GEA
55.7
EUR
SIAL655244-1G
SIAL655244-5G
2-Chloro-3-methoxybenzaldehyde, Sigma-Aldrich®
2-Chloro-3-methoxybenzaldehyde
Formula:
C₈H₇ClO₂ MW: 170.5930 g/mol |
MDL Number:
MFCD07369787 CAS Number: 54881-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...