Benzyl isovalerate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W215201-10KG-K W215201-1KG-K W215201-5KG-K
SIALW215201-10KG-KEA 807 EUR
SIALW215201-10KG-K SIALW215201-1KG-K SIALW215201-5KG-K
Benzyl isovalerate, Sigma-Aldrich®
Benzyl isovalerate
Formula: C₁₂H₁₆O₂
MDL Number: MFCD00048348
CAS Number: 103-38-8
EINECS: 203-106-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR