Propylphosphonic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL305685-1GEA 95.5 EUR
SIAL305685-1G
Propylphosphonic acid, Sigma-Aldrich®
Propylphosphonic acid
Formula: C₃H₉O₃P
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR