Nickel(II) carbonate (basic) hydrate (≥40% Ni) ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Danger
40106.30 40106.A1
40106.30EA 68.9 EUR
40106.30 40106.A1
Nickel(II) carbonate (basic) hydrate (≥40% Ni) ≥99.5% (metals basis)
Nickel(II) carbonate (basic) hydrate
Purity: 99.5% CAS No.: 155775-31-8 Molecular Formula: CNiO3.2H2NiO2.xH2O Formula Weight: 304.18(anhy) Physical Form: Powder Appearance: Green Odour: Odourless UN No.: 3077 MDL No.: MFCD00016262
Nickel plating pH adjuster, catalysts, ceramic Colours and glazes
Formula: CNiO₃·2H₂NiO₂·xH₂O
MW: 304.18 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00016262
CAS Number: 155775-31-8
EINECS: 240-408-8
UN: 3077
ADR: 9 III