Fenvalerate

Supplier: EHRENSTORFER GMBH

EHERC13630000EA 95.5 EUR
EHERC13630000
Fenvalerate
Fenvalerate
Formula: C₂₅H₂₂ClNO₃
CAS Number: 51630-58-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR