Fenvalerate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALF1428-25MGEA 127 EUR
SIALF1428-25MG
Fenvalerate, Sigma-Aldrich®
Fenvalerate
Formula: C₂₅H₂₂ClNO₃
CAS Number: 51630-58-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR