1,2,3,4-Tetrahydroisoquinoline hydrochloride

Supplier: Thermo Scientific

BTB09216.1 BTB09216.10
MAYBBTB09216.1EA 0 EUR
MAYBBTB09216.1 MAYBBTB09216.10
1,2,3,4-Tetrahydroisoquinoline hydrochloride
1,2,3,4-Tetrahydroisoquinoline hydrochloride
Formula: C₉H₁₂ClN
CAS Number: 14099-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR