2,6-Dibromoquinone-4-chloroimide 97%
Supplier: Thermo Scientific
Danger
Synonyms: 2,6-Dibromo-N-chloro-p-benzoquinoneimine
406470050 406470250
ACRO406470050EA 164 EUR
ACRO406470050 ACRO406470250
2,6-Dibromoquinone-4-chloroimide 97%
2,6-Dibromoquinone-4-chloroimide
Formula: C₆H₂Br₂ClNO
Melting Pt: 76…83 °C
Density: 1.81 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00013792
CAS Number: 537-45-1
EINECS: 208-667-1
UN: 3224
ADR: 4.1,
Order Now
Specification Test Results
Appearance Yellow to ochre Powder or crystalline needles (May darken on storage)
Infrared spectrum Conforms
Melting point 76 to 83 °C
Titration Iodometric ≥96.0 %