Orotic acid
Supplier: Thermo Scientific
Synonyms: 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
SB00661.1 SB00661.10 SB00661.50
MAYBSB00661.1EA 0 EUR
MAYBSB00661.1 MAYBSB00661.10 MAYBSB00661.50
Orotic acid
Orotic acid
Formula: C₅H₄N₂O₄
MDL Number: MFCD00006027
CAS Number: 65-86-1
EINECS: 200-619-8