Ethyl (ethoxymethylene)cyanoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ethyl 2-Cyano-3-Ethoxyacrylate
M-Clarity™ quality level = MQ100
E28005-100G
E28005-25G
SIALE28005-100GEA
45.3
EUR
SIALE28005-100G
SIALE28005-25G
Ethyl (ethoxymethylene)cyanoacetate, Sigma-Aldrich®
Ethyl (ethoxymethylene)cyanoacetate
Formula:
C₈H₁₁NO₃ MW: 169.18 g/mol |
MDL Number:
MFCD00009136 CAS Number: 94-05-3 EINECS: 202-299-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...