N-(2-Chloroethyl)acetamide 98%
Supplier: Thermo Fisher Scientific
Warning
184520100 184520500
ACRO184520100EA 43.3 EUR
ACRO184520100 ACRO184520500
N-(2-Chloroethyl)acetamide 98%
N-(2-Chloroethyl)acetamide
Formula: C₄H₈ClNO
Density: 1.2 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000965
CAS Number: 7355-58-0
EINECS: 230-884-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %