3-(Trimethoxysilyl)propyl methacrylate, Sigma-Aldrich®

Supplier: Merck

Synonyms: γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane

M-Clarity™ quality level = MQ200

440159-100ML 440159-500ML
SIAL440159-100MLEA 63.1 EUR
SIAL440159-100ML SIAL440159-500ML
3-(Trimethoxysilyl)propyl methacrylate, Sigma-Aldrich®
3-(Trimethoxysilyl)propyl methacrylate
Formula: C₁₀H₂₀O₅Si
MW: 248.35 g/mol
MDL Number: MFCD00008593
CAS Number: 2530-85-0
EINECS: 219-785-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR