4-(Methoxycarbonyl)phenylboronic acid
Supplier: Thermo Scientific
Synonyms:
(4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid
AC35956.250
AC35956.1
MAYBAC35956.250EA
0
EUR
MAYBAC35956.250
MAYBAC35956.1
4-(Methoxycarbonyl)phenylboronic acid
4-(Methoxycarbonyl)phenylboronic acid
Formula:
C₈H₉BO₄ MW: 179.97 g/mol |
MDL Number:
MFCD01632203 CAS Number: 99768-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...