4-(Benzyloxy)-3-methoxybenzaldehyde

Supplier: Thermo Scientific

BTB07524.1 BTB07524EA BTB07524EE
MAYBBTB07524.1EA 0 EUR
MAYBBTB07524.1 MAYBBTB07524EA MAYBBTB07524EE
4-(Benzyloxy)-3-methoxybenzaldehyde
4-(Benzyloxy)-3-methoxybenzaldehyde
Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00003365
CAS Number: 2426-87-1
EINECS: 219-379-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR