N,N,N',N'-Tetramethylbenzidine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N,n,n,n-tetramethylbenzidine, 4,4'-Bis(dimethylamino)biphenyl
M-Clarity™ quality level = MQ100
T5513-25G
T5513-5G
SIALT5513-25GEA
476
EUR
SIALT5513-25G
SIALT5513-5G
N,N,N',N'-Tetramethylbenzidine, Sigma-Aldrich®
N,N,N',N'-Tetramethylbenzidine
Formula:
C₁₆H₂₀N₂ |
MDL Number:
MFCD00008310 CAS Number: 366-29-0 EINECS: 206-676-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...