1,8-Bis(dimethylamino)naphthalene, Sigma-Aldrich®
Supplier: Merck
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
M-Clarity™ quality level = MQ200
158496-10G 158496-50G
SIAL158496-10GEA 51.8 EUR
SIAL158496-10G SIAL158496-50G
1,8-Bis(dimethylamino)naphthalene, Sigma-Aldrich®
1,8-Bis(dimethylamino)naphthalene
Formula: C₁₄H₁₈N₂
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6
UN: 3263
ADR: 8,III