3,6-Dimethyl-1,4-dioxane-2,5-dione, Sigma-Aldrich®
Supplier: Merck
Synonyms: DL-Lactide
M-Clarity™ quality level = MQ200
303143-100G 303143-25G
SIAL303143-100GEA 200 EUR
SIAL303143-100G SIAL303143-25G
3,6-Dimethyl-1,4-dioxane-2,5-dione, Sigma-Aldrich®
3,6-Dimethyl-1,4-dioxane-2,5-dione
Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00011685
CAS Number: 95-96-5
EINECS: 202-468-3