Vitamin K1 (phytomenadione, phylloquinone)
Supplier: Roth Carl
Synonyms: 2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
3804.1 3804.2
ROTH3804.1EA 0 EUR
ROTH3804.1 ROTH3804.2
Vitamin K1 (phytomenadione, phylloquinone)
Vitamin K1 (Phytomenadione, Phylloquinone)
Formula: C₃₁H₄₆O₂
MW: 450.6958 g/mol
MDL Number: MFCD00214063
CAS Number: 84-80-0
EINECS: 201-564-2