Vitamin K1 (phytomenadione, phylloquinone), Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
MQ100
V3501-1G
V3501-10G
SIALV3501-1GEA
163
EUR
SIALV3501-1G
SIALV3501-10G
Vitamin K1 (phytomenadione, phylloquinone), Sigma-Aldrich®
Vitamin K1 (Phytomenadione, Phylloquinone)
Formula:
C₃₁H₄₆O₂ MW: 450.6958 g/mol |
MDL Number:
MFCD00214063 CAS Number: 84-80-0 EINECS: 201-564-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...