1-Vinyl-2-pyrrolidone stabilised, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone

Stabilization: Stabilised with sodium hydroxide
M-Clarity™ quality level = MQ200

V3409-18KG V3409-1KG V3409-250G V3409-5G
SIALV3409-18KGEA 1820 EUR
SIALV3409-18KG SIALV3409-1KG SIALV3409-250G SIALV3409-5G
1-Vinyl-2-pyrrolidone stabilised, Sigma-Aldrich®
1-Vinyl-2-pyrrolidone
Formula: C₆H₉NO
MW: 111.1418 g/mol
MDL Number: MFCD00003197
CAS Number: 88-12-0
EINECS: 201-800-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR