Cyanamide Stabilised, Sigma-Aldrich®
Supplier: Merck
MQ200
187364-100G
187364-250G
187364-25G
187364-5G
SIAL187364-100GEA
110
EUR
SIAL187364-100G
SIAL187364-250G
SIAL187364-25G
SIAL187364-5G
Cyanamide Stabilised, Sigma-Aldrich®
Cyanamide
Formula:
CH₂N₂ MW: 42.04 g/mol |
MDL Number:
MFCD00007572 CAS Number: 420-04-2 EINECS: 206-992-3 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...