3-Thienylacetic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: Thiophene-3-acetic acid, 3-Thiopheneacetic acid

M-Clarity™ quality level = MQ100

220639-10G 220639-50G
SIAL220639-10GEA 106 EUR
SIAL220639-10G SIAL220639-50G
3-Thienylacetic acid, Sigma-Aldrich®
3-Thienylacetic acid
Formula: C₆H₆O₂S
MDL Number: MFCD00005473
CAS Number: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR