2-Chloro-6-(tributylstannyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Danger

719609-500MG 719609-1G
SIAL719609-500MGEA 121 EUR
SIAL719609-500MG SIAL719609-1G
2-Chloro-6-(tributylstannyl)pyridine, Sigma-Aldrich®
2-Chloro-6-(tributylstannyl)pyridine
Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD10699155
CAS Number: 263698-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR